Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2,4,5-Cyclohexanetetracarboxylic acid 97%

Supplier: AMBEED, INC
Warning

A103160-1G A103160-5G A103160-10G A103160-250MG A103160-25G A103160-1G
103796-494EA 6 USD InStock
103796-494 103822-198 77008-440 77008-442 77008-444 77309-070 77309-072
1,2,4,5-Cyclohexanetetracarboxylic acid 97%
1,2,4,5-Cyclohexanetetracarboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₈
Melting Pt: >220 °C
Storage Temperature: Ambient
MDL Number: MFCD00435556
CAS Number: 15383-49-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty