Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4H-Cyclopenta[1,2-b:5,4-b']dithiophen-4-one 98%

Supplier: AMBEED, INC
Warning

A182390-1G A182390-5G A182390-250MG A182390-25G A182390-1G A182390-5G A182390-100MG
103807-706EA 31 USD InStock
103807-706 103846-078 77029-490 77029-492 77083-620 77301-780 77667-516
4H-Cyclopenta[1,2-b:5,4-b']dithiophen-4-one 98%
4H-Cyclopenta[1,2-b:5,4-b']dithiophen-4-one
Precaución: for Research Use Only


Formula: C₉H₄OS₂
Storage Temperature: Ambient
MDL Number: MFCD16619150
CAS Number: 25796-77-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty