Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclopentane-1,3-dicarboxylic acid 95%

Supplier: AMBEED, INC
Warning

A127545-1G A127545-1G A127545-5G A127545-10G A127545-25G
103800-974EA 17 USD InStock
103800-974 77313-214 77313-216 77871-580 77871-581
Cyclopentane-1,3-dicarboxylic acid 95%
Cyclopentane-1,3-dicarboxylic acid
Precaución: for Research Use Only


Formula: C₇H₁₀O₄
MW: 158.15 g/mol
Boiling Pt: 320.7 °C (760 mm Hg)
Flash Pt: 161.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01735467
CAS Number: 4056-78-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty