Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclopentylmethanol 98%

Supplier: AMBEED, INC
Warning

A141416-1G A141416-10G A141416-1G A141416-25G A141416-5G
103802-992EA 5 USD InStock
103802-992 76689-020 77316-150 77316-152 77316-154
Cyclopentylmethanol 98%
Cyclopentylmethanol
Precaución: for Research Use Only


Formula: C₆H₁₂O
MW: 100.16
Boiling Pt: 163 °C (760 mm Hg)
Density: 0.926
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00001384
CAS Number: 3637-61-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty