Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclopentylboronic acid 97%

Supplier: AMBEED, INC
Warning

A163434-1G A163434-5G A163434-100G A163434-10G A163434-1G A163434-5G
103805-726EA 5 USD InStock
103805-726 103843-986 76985-158 76985-160 77300-658 77320-084
Cyclopentylboronic acid 97%
Cyclopentylboronic acid
Precaución: for Research Use Only


Formula: C₅H₁₁BO₂
Boiling Pt: 235 °C (760 mm Hg)
Melting Pt: 126 °C
Flash Pt: 95.9 °C
Storage Temperature: Freezer
MDL Number: MFCD01074541
CAS Number: 63076-51-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty