Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1-Cyclopropanedicarboxylic acid 97%

Supplier: AMBEED, INC
Warning

A107915-5G A107915-100G A107915-1G A107915-25G A107915-500G A107915-5G
103823-038EA 9 USD InStock
103823-038 77008-734 77008-736 77008-738 77008-740 77309-884
1,1-Cyclopropanedicarboxylic acid 97%
1,1-Cyclopropanedicarboxylic acid
Precaución: for Research Use Only


Formula: C₅H₆O₄
MW: 130.1
Boiling Pt: 371
Melting Pt: 130 °C
Flash Pt: 192.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00013727
CAS Number: 598-10-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty