Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclopropanecarboxamidine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A264831-1G A264831-5G A264831-100G A264831-10G A264831-500G A264831-1G A264831-25G
103812-694EA 9 USD InStock
103812-694 103852-588 76827-186 76827-188 76827-190 77092-254 77304-904
Cyclopropanecarboxamidine hydrochloride 97%
Cyclopropanecarboxamidine hydrochloride
Precaución: for Research Use Only


Formula: C₄H₈N₂
Boiling Pt: 133.6 °C (760 mm Hg)
Melting Pt: 123 °C
Storage Temperature: Ambient
MDL Number: MFCD00053010
CAS Number: 57297-29-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty