Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cytosine-β-D-arabinofuranoside hydrochloride 97%

Supplier: AMBEED, INC
Warning

A101511-1G A101511-5G A101511-100MG A101511-10G A101511-25MG A101511-1G A101511-5G A101511-25G
103796-008EA 9 USD InStock
103796-008 103821-690 76687-040 76687-042 76687-044 77297-138 77297-140 77878-795
Cytosine-β-D-arabinofuranoside hydrochloride 97%
Cytosine-beta-D-arabinofuranoside hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₄ClN₃O₅
MW: 279.677 g/mol
Boiling Pt: 545.7 °C (760 mm Hg)
Melting Pt: 197…198 °C
Flash Pt: 283.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012839
CAS Number: 69-74-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty