Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-Cycloserine 98%

Supplier: AMBEED, INC
Warning

A170761-5G A170761-25G A170761-5G A170761-100MG A170761-10G A170761-1G
103844-814EA 9 USD InStock
103844-814 77081-928 77081-930 77286-836 77286-838 77286-840
D-Cycloserine 98%
D-Cycloserine
Precaución: For research use only.


Formula: C₃H₆N₂O₂
MW: 102.09 g/mol
Melting Pt: 147 °C
Storage Temperature: Freezer
MDL Number: MFCD00005353
CAS Number: 68-41-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty