Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-(-)-Glutamine 96%

Supplier: AMBEED, INC
Warning

A259333-5G A259333-100G A259333-100MG A259333-10G A259333-1MG A259333-50MG A259333-25G A259333-5G
103852-312EA 18 USD InStock
103852-312 76988-156 76988-158 76988-160 76988-162 76988-164 77091-858 77091-860
D-(-)-Glutamine 96%
D-(-)-Glutamine
Precaución: for Research Use Only


Formula: C₅H₁₀N₂O₃
MW: 146.15
Boiling Pt: 445.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00065607
CAS Number: 5959-95-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty