Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Di(1H-imidazol-1-yl)methane 97%

Supplier: AMBEED, INC
Warning

A165280-1G A165280-5G A165280-100MG A165280-250MG A165280-1G
103805-952EA 55 USD InStock
103805-952 103844-210 77163-332 77163-334 77320-412
Di(1H-imidazol-1-yl)methane 97%
Di(1H-imidazol-1-yl)methane
Precaución: Inert atmosphere, room temperature


Formula: C₇H₈N₄
MW: 148.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00159695
CAS Number: 84661-56-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty