Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1'-Carbonimidoylbis-1H-imidazole 95%

Supplier: AMBEED, INC
Warning

A110025-1G A110025-5G A110025-1G A110025-5G
103797-604EA 21 USD InStock
103797-604 103823-412 77310-218 77310-220
1,1'-Carbonimidoylbis-1H-imidazole 95%
1,1'-Carbonimidoylbis-1H-imidazole
Precaución: for Research Use Only


Formula: C₇H₇N₅
Boiling Pt: 355.1 °C (760 mm Hg)
Melting Pt: 103…104 °C
Flash Pt: 169 °C
Storage Temperature: Freezer
MDL Number: MFCD10699388
CAS Number: 104619-51-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty