Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2'-Dipyrrolylmethane 97%

Supplier: AMBEED, INC
Warning

A197551-100MG A197551-250MG A197551-1G A197551-25G
103816-934EA 29 USD InStock
103816-934 76873-270 76873-272 77085-566 77302-554
2,2'-Dipyrrolylmethane 97%
2,2'-Dipyrrolylmethane
Precaución: for Research Use Only


Formula: C₉H₁₀N₂
Boiling Pt: 317.8 °C (760 mm Hg)
Flash Pt: 142.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03931560
CAS Number: 21211-65-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty