Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dibenzylamine 98%

Supplier: AMBEED, INC
Danger

A281081-100G A281081-25G
103809-938EA 32 USD InStock
103809-938 76802-180 76802-182 76837-184
Dibenzylamine 98%
Dibenzylamine
Precaución: for Research Use Only


Formula: C₆H₅CH₂NHCH₂C₆H₅
MW: 197.28 g/mol
Boiling Pt: 300
Melting Pt: -26
Flash Pt: 143 °C
Storage Temperature: Ambient
MDL Number: MFCD00004770
CAS Number: 103-49-1
UN: 2735
ADR: 8 Ⅲ
Merck Index: 13,03035
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty