Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Dicyclohexylphosphino-2',6'-diisopropoxybiphenyl 97%

Supplier: AMBEED, INC
Warning

A125288-1G A125288-100G A125288-10G A125288-250MG A125288-25G A125288-500G A125288-1G A125288-5G
103800-662EA 11 USD InStock
103800-662 77258-242 77258-244 77258-246 77258-248 77258-250 77312-814 77312-816
2-Dicyclohexylphosphino-2',6'-diisopropoxybiphenyl 97%
2-Dicyclohexylphosphino-2',6'-diisopropoxybiphenyl
Precaución: for Research Use Only


Formula: C₃₀H₄₃O₂P
Melting Pt: 123…126 °C
Flash Pt: 360.7 °C
Storage Temperature: Ambient
MDL Number: MFCD06798294
CAS Number: 787618-22-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty