Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl aminomalonate hydrochloride 98%

Supplier: AMBEED, INC
Warning

A306980-5G A306980-100G A306980-500G A306980-1000G A306980-25G A306980-5G
103810-044EA 53 USD InStock
103810-044 103835-664 77277-436 77277-438 77305-940 77305-942 77305-944
Diethyl aminomalonate hydrochloride 98%
Diethyl aminomalonate hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₄ClNO₄
MW: 211.65 g/mol
Melting Pt: 165…170 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012510
CAS Number: 13433-00-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty