Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl-2-ethoxymalonate 97%

Supplier: AMBEED, INC
Warning

A140176-1G A140176-250MG A140176-1G A140176-5G
103802-840EA 69 USD InStock
103802-840 77027-478 77315-954 77315-956
Diethyl-2-ethoxymalonate 97%
Diethyl-2-ethoxymalonate
Precaución: for Research Use Only


Formula: C₉H₁₆O₅
Boiling Pt: 260.8 °C (760 mm Hg)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00087125
CAS Number: 37555-99-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty