Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl ethylidenemalonate 97%

Supplier: AMBEED, INC
Warning

A105377-1G A105377-5G A105377-100G A105377-10G A105377-25G A105377-1G A105377-5G
103796-880EA 5 USD InStock
103796-880 103822-608 77245-392 77245-394 77245-396 77283-352 77309-480
Diethyl ethylidenemalonate 97%
Diethyl ethylidenemalonate
Precaución: for Research Use Only


Formula: C₉H₁₄O₄
MW: 186.21 g/mol
Boiling Pt: 232.1 °C (760 mm Hg)
Flash Pt: 85.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00009145
CAS Number: 1462-12-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty