Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl mesoxalate (diethyl ketomalonate) 95%

Supplier: AMBEED, INC
Warning

A145955-1G A145955-5G A145955-100G A145955-10G A145955-500G A145955-1G A145955-25G A145955-5G
103803-608EA 5 USD InStock
103803-608 103841-692 77110-046 77110-048 77110-050 77316-996 77316-998 77317-000
Diethyl mesoxalate (diethyl ketomalonate) 95%
Diethyl mesoxalate (diethyl ketomalonate)
Precaución: for Research Use Only


Formula: C₇H₁₀O₅
MW: 174.15 g/mol
Boiling Pt: 209 °C (760 mm Hg)
Flash Pt: 90.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00009121
CAS Number: 609-09-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty