Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl ethylmalonate 98%

Supplier: AMBEED, INC
Warning

A202766-5G A202766-1000G A202766-100G A202766-500G A202766-25G A202766-5G
103848-060EA 5 USD InStock
103848-060 76999-408 76999-410 76999-412 77302-826 77302-828
Diethyl ethylmalonate 98%
Diethyl ethylmalonate
Precaución: for Research Use Only


Formula: C₉H₁₆O₄
MW: 188.22 g/mol
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00009167
CAS Number: 133-13-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty