Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl isopropylmalonate 98%

Supplier: AMBEED, INC
Warning

A683609-5G A683609-100G A683609-10G A683609-500G A683609-25G
103840-546EA 7 USD InStock
103840-546 76697-222 76697-224 76697-226 76844-750
Diethyl isopropylmalonate 98%
Diethyl isopropylmalonate
Precaución: for Research Use Only


Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
Boiling Pt: 62 °C at 0.35 mmHg
Flash Pt: 96.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00040491
CAS Number: 759-36-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty