Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl malonyldicarbamate 97%

Supplier: AMBEED, INC
Warning

A322832-1G A322832-5G A322832-100G A322832-10G A322832-25G
103825-310EA 8 USD InStock
103825-310 103836-248 76839-166 76839-168 76875-884 76875-886 77306-278
Diethyl malonyldicarbamate 97%
Diethyl malonyldicarbamate
Precaución: for Research Use Only


Formula: C₉H₁₄N₂O₆
MW: 246.22 g/mol
Melting Pt: 124 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00098229
CAS Number: 49754-15-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty