Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(-1,5-Bis(5-nitrofuran-2-yl)penta-1,4-dien-3-ylidene)hydrazinecarboximidamide hydrochloride 97%

Supplier: AMBEED, INC
Warning

A129165-1G A129165-250MG A129165-1G
103801-236EA 56 USD InStock
103801-236 77026-972 77298-332
2-(-1,5-Bis(5-nitrofuran-2-yl)penta-1,4-dien-3-ylidene)hydrazinecarboximidamide hydrochloride 97%
2-(-1,5-Bis(5-nitrofuran-2-yl)penta-1,4-dien-3-ylidene)hydrazinecarboximidamide hydrochloride
Precaución: for Research Use Only


Formula: C₁₄H₁₃ClN₆O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00242893
CAS Number: 2315-20-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty