Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diisopropyl carbonate 98%

Supplier: AMBEED, INC
Warning

A118846-1G A118846-10G A118846-250MG A118846-25G A118846-5G A118846-1G
103799-116EA 15 USD InStock
103799-116 76797-250 76797-252 76797-254 77297-628 77311-790
Diisopropyl carbonate 98%
Diisopropyl carbonate
Precaución: for Research Use Only


Formula: C₇H₁₄O₃
Boiling Pt: 147 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07779375
CAS Number: 6482-34-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty