Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl 3-aminopentanedioate hydrochloride 97%

Supplier: AMBEED, INC
Warning

A127385-1G A127385-100MG A127385-250MG
103800-956EA 66 USD InStock
103800-956 77009-738 77009-740
Dimethyl 3-aminopentanedioate hydrochloride 97%
Dimethyl 3-aminopentanedioate hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₄ClNO₄
MW: 211.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14705045
CAS Number: 77313-10-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty