Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl 4-fluorophthalate 95%

Supplier: AMBEED, INC
Warning

A196364-5G A196364-1G A196364-250MG
103847-450EA 111 USD InStock
103847-450 76800-192 76800-194
Dimethyl 4-fluorophthalate 95%
Dimethyl 4-fluorophthalate
Precaución: for Research Use Only


Formula: C₁₀H₉FO₄
Boiling Pt: 251.994 °C (760 mm Hg)
Flash Pt: 103.127 °C
Storage Temperature: Ambient
MDL Number: MFCD09263469
CAS Number: 110706-50-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty