Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl 5-fluoroisophthalate 97%

Supplier: AMBEED, INC
Warning

A373425-5G A373425-1G A373425-250MG
103826-532EA 18 USD InStock
103826-532 103837-550 76840-816 77003-540
Dimethyl 5-fluoroisophthalate 97%
Dimethyl 5-fluoroisophthalate
Precaución: for Research Use Only


Formula: C₁₀H₉FO₄
Boiling Pt: 288.3 °C (760 mm Hg)
Flash Pt: 124.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00230549
CAS Number: 17449-48-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty