Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl 5-methylpyridine-2,3-dicarboxylate 97%

Supplier: AMBEED, INC
Warning

A323719-1G A323719-5G A323719-100G A323719-25G
103825-354EA 9 USD InStock
103825-354 103836-296 76839-208 76839-210 77194-876 77306-312
Dimethyl 5-methylpyridine-2,3-dicarboxylate 97%
Dimethyl 5-methylpyridine-2,3-dicarboxylate
Precaución: for Research Use Only


Formula: C₁₀H₁₁NO₄
Boiling Pt: 292.2 °C (760 mm Hg)
Flash Pt: 130.5 °C
Storage Temperature: Ambient
MDL Number: MFCD05662424
CAS Number: 112110-16-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty