Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl-2,6-pyridinedicarboxylate 98%

Supplier: AMBEED, INC
Warning

A149651-5G A149651-100G A149651-10G A149651-25G A149651-5G A149651-500G
103842-190EA 5 USD InStock
103842-190 76798-572 76798-574 77317-690 77317-692 77624-594
Dimethyl-2,6-pyridinedicarboxylate 98%
Dimethyl 2,6-Pyridinedicarboxylate
Precaución: for Research Use Only


Formula: C₉H₉NO₄
Boiling Pt: 321.7 °C (760 mm Hg)
Melting Pt: 121…125 °C
Flash Pt: 148.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00134493
CAS Number: 5453-67-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty