Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diphenyl-2-pyridylphosphine 98%

Supplier: AMBEED, INC
Warning

A101214-1G A101214-5G A101214-10G A101214-25G A101214-500G A101214-1G A101214-5G A101214-1000G
103795-860EA 5 USD InStock
103795-860 103821-544 76796-266 76796-268 76796-270 77282-682 77297-022 77878-785
Diphenyl-2-pyridylphosphine 98%
Diphenyl-2-pyridylphosphine
Precaución: for Research Use Only


Formula: C₁₇H₁₄NP
Boiling Pt: 377.5 °C (760 mm Hg)
Melting Pt: 82…84 °C
Flash Pt: 182.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00192108
CAS Number: 37943-90-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty