Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Di-tert-butyl-N,N-diisopropylphosphoramidite 95%

Supplier: AMBEED, INC
Warning

A104095-1G A104095-5G A104095-10G A104095-25G A104095-1G A104095-5G
103796-654EA 6 USD InStock
103796-654 103822-366 76796-402 76796-404 77309-224 77309-226
Di-tert-butyl-N,N-diisopropylphosphoramidite 95%
Di-tert-butyl-N,N-diisopropylphosphoramidite
Precaución: for Research Use Only


Formula: C₁₄H₃₂NO₂P
MW: 277.39 g/mol
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00153506
CAS Number: 137348-86-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty