Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

DL-Glutamine 97%

Supplier: AMBEED, INC
Warning

A228319-5G A228319-100G A228319-500G A228319-25G A228319-5G
103850-296EA 21 USD InStock
103850-296 76987-226 76987-228 77303-828 77303-830
DL-Glutamine 97%
DL-Glutamine
Precaución: for Research Use Only


Formula: C₅H₁₀N₂O₃
Boiling Pt: 445.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00065103
CAS Number: 6899-04-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty