Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D(+)-Ribonic acid γ-lactone 95%

Supplier: AMBEED, INC
Warning

A648099-5G A648099-100MG A648099-10G A648099-250MG A648099-25G A648099-5G
103840-386EA 80 USD InStock
103840-386 76775-542 76775-544 76775-546 76775-548 77308-058
D(+)-Ribonic acid γ-lactone 95%
D(+)-Ribonic acid γ-lactone
Precaución: for Research Use Only


Formula: C₅H₈O₅
Boiling Pt: 364.3 °C (760 mm Hg)
Melting Pt: 83…85 °C
Flash Pt: 162.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00063241
CAS Number: 5336-08-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty