Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethoxydiphenylphosphine 98%

Supplier: AMBEED, INC

A178604-5G A178604-100G A178604-10G A178604-1G A178604-500G A178604-25G A178604-5G
103845-696EA 7 USD InStock
103845-696 76799-624 76799-626 76799-628 76799-630 77301-566 77301-568
Ethoxydiphenylphosphine 98%
Ethoxydiphenylphosphine
Precaución: for Research Use Only


Formula: C₁₄H₁₅OP
MDL Number: MFCD00009087
CAS Number: 719-80-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty