Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 1-aminocyclohexanecarboxylate hydrochloride 97%

Supplier: AMBEED, INC
Warning

A101085-1G A101085-250MG A101085-1G
103795-798EA 82 USD InStock
103795-798 77160-192 77296-984
Ethyl 1-aminocyclohexanecarboxylate hydrochloride 97%
Ethyl 1-aminocyclohexanecarboxylate hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD07366916
CAS Number: 63203-48-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty