Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 1H-imidazole-4-carboxylate 97%

Supplier: AMBEED, INC
Warning

A250609-1G A250609-5G A250609-1G A250609-25G A250609-5G A250609-100G A250609-10G A250609-250MG A250609-500G
103811-934EA 6 USD InStock
103811-934 103851-772 77091-122 77091-124 77091-126 77098-678 77098-680 77098-682 77685-712
Ethyl 1H-imidazole-4-carboxylate 97%
Ethyl 1H-imidazole-4-carboxylate
Precaución: for Research Use Only


Formula: C₆H₈N₂O₂
Boiling Pt: 319.5 °C (760 mm Hg)
Melting Pt: 160…162 °C
Flash Pt: 147 °C
Storage Temperature: Ambient
MDL Number: MFCD02646639
CAS Number: 23785-21-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty