Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl pyrazole-3-carboxylate 97%

Supplier: AMBEED, INC
Warning

A328993-1G A328993-5G A328993-25G A328993-100G A328993-10G A328993-500G A328993-1G A328993-5G
103825-504EA 5 USD InStock
103825-504 103836-458 76839-426 77239-720 77239-722 77239-724 77306-380 77306-382
Ethyl pyrazole-3-carboxylate 97%
Ethyl pyrazole-3-carboxylate
Precaución: for Research Use Only


Formula: C₆H₈N₂O₂
Boiling Pt: 279.2 °C (760 mm Hg)
Melting Pt: 279.2 °C
Flash Pt: 123 °C
Storage Temperature: Ambient
MDL Number: MFCD00159643
CAS Number: 5932-27-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty