Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 1H-pyrrolo[3,4-c]pyridine-2(3H)-carboxylate 95%

Supplier: AMBEED, INC
Warning

A132860-1G A132860-100MG A132860-250MG
103801-798EA 147 USD InStock
103801-798 77027-146 77027-148
Ethyl 1H-pyrrolo[3,4-c]pyridine-2(3H)-carboxylate 95%
Ethyl 1H-pyrrolo[3,4-c]pyridine-2(3H)-carboxylate
Precaución: for Research Use Only


Formula: C₁₀H₁₂N₂O₂
Boiling Pt: 306.5 °C (760 mm Hg)
Flash Pt: 139.2 °C
Storage Temperature: Ambient
MDL Number: MFCD13183761
CAS Number: 147740-04-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty