Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Methoxyphenyl)acetic acid ethyl ester 98%

Supplier: AMBEED, INC
Danger

A754029-5G A754029-25G A754029-5G A754029-10G
103840-860EA 8 USD InStock
103840-860 76845-092 76845-094 77158-900
(4-Methoxyphenyl)acetic acid ethyl ester 98%
(4-Methoxyphenyl)acetic acid ethyl ester
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₃
Boiling Pt: 272 °C (760 mm Hg)
Flash Pt: 108.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00040760
CAS Number: 14062-18-1
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty