Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-(6-aminopyridin-3-yl)acetate 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A178254-1G A178254-1G A178254-250MG A178254-100MG
103807-306EA 315 USD InStock
103807-306 77083-052 77163-788 77683-427
Ethyl 2-(6-aminopyridin-3-yl)acetate 95%, Ambeed.Inc
Ethyl 2-(6-aminopyridin-3-yl)acetate
Precaución: For research use only.


Formula: C₉H₁₂N₂O₂
MW: 180.2 g/mol
Boiling Pt: 312.9 °C (760 mm Hg)
Flash Pt: 143 °C
Storage Temperature: Refrigerator
MDL Number: MFCD20703180
CAS Number: 1174229-72-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty