Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Pyridyl)acetic acid ethyl ester 98%

Supplier: AMBEED, INC
Warning

A183259-1G A183259-100G A183259-10G A183259-25G A183259-1G A183259-5G
103815-624EA 5 USD InStock
103815-624 103846-186 76799-804 76799-806 76799-808 77083-780 77083-782
(3-Pyridyl)acetic acid ethyl ester 98%
(3-Pyridyl)acetic acid ethyl ester
Precaución: for Research Use Only


Formula: C₉H₁₁NO₂
Boiling Pt: 274.4 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006411
CAS Number: 39931-77-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty