Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-amino-1H-imidazole-5-carboxylate 97%

Supplier: AMBEED, INC
Warning

A127539-1G A127539-100MG A127539-250MG A127539-1G A127539-5G
103800-972EA 41 USD InStock
103800-972 76996-164 76996-166 77298-222 77298-224
Ethyl-2-amino-1H-imidazole-5-carboxylate 97%
Ethyl-2-amino-1H-imidazole-5-carboxylate
Precaución: for Research Use Only


Formula: C₆H₉N₃O₂
MW: 155.1546 g/mol
Boiling Pt: 363.6 °C (760 mm Hg)
Flash Pt: 173.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09864446
CAS Number: 149520-94-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty