Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-aminobenzo[d]thiazole-6-carboxylate 98%

Supplier: AMBEED, INC
Warning

A138105-1G A138105-100G A138105-1G A138105-25G A138105-5G A138105-10G
103802-554EA 5 USD InStock
103802-554 77095-300 77315-568 77315-570 77315-572 77685-185
Ethyl-2-aminobenzo[d]thiazole-6-carboxylate 98%
Ethyl-2-aminobenzo[d]thiazole-6-carboxylate
Precaución: for Research Use Only


Formula: C₁₀H₁₀N₂O₂S
MW: 222.26 g/mol
Boiling Pt: 386.8 °C (760 mm Hg)
Melting Pt: 241…243 °C
Flash Pt: 187.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00102724
CAS Number: 50850-93-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty