Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-aminothiophene-3-carboxylate 97%

Supplier: AMBEED, INC
Warning

A187547-5G A187547-25G A187547-100G A187547-10G
103816-026EA 9 USD InStock
103816-026 103846-616 77084-316 77164-124 77164-126
Ethyl-2-aminothiophene-3-carboxylate 97%
Ethyl-2-aminothiophene-3-carboxylate
Precaución: for Research Use Only


Formula: C₇H₉NO₂S
Boiling Pt: 273.6 °C (760 mm Hg)
Melting Pt: 42 °C
Flash Pt: 119.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01566303
CAS Number: 31891-06-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty