Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-chlorothiazole-4-carboxylate 95%

Supplier: AMBEED, INC
Warning

A271271-1G A271271-5G A271271-100G A271271-10G A271271-250MG A271271-1G A271271-25G
103813-042EA 20 USD InStock
103813-042 103852-952 76787-188 76787-190 76787-192 77092-732 77305-070
Ethyl-2-chlorothiazole-4-carboxylate 95%
Ethyl-2-chlorothiazole-4-carboxylate
Precaución: for Research Use Only


Formula: C₆H₆ClNO₂S
MW: 191.64 g/mol
Boiling Pt: 283.9 °C (760 mm Hg)
Melting Pt: 75…79 °C
Flash Pt: 125.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11045348
CAS Number: 41731-52-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty