Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-cyano-4,4-diethoxybutyrate 97%

Supplier: AMBEED, INC
Warning

A331124-5G A331124-100G A331124-10G A331124-500G A331124-1G A331124-25G
103825-572EA 7 USD InStock
103825-572 103836-536 76803-062 76803-064 76803-066 76839-520 76839-522
Ethyl-2-cyano-4,4-diethoxybutyrate 97%
Ethyl-2-cyano-4,4-diethoxybutyrate
Precaución: for Research Use Only


Formula: C₁₁H₁₉NO₄
Boiling Pt: 335.2 °C (760 mm Hg)
Flash Pt: 146.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02094250
CAS Number: 52133-67-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty