Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl ethoxyiminoacetate 95%

Supplier: AMBEED, INC
Warning

A127074-1G A127074-100MG A127074-250MG A127074-10G
103800-910EA 26 USD InStock
103800-910 77026-874 77026-876 77670-794
Ethyl ethoxyiminoacetate 95%
Ethyl ethoxyiminoacetate
Precaución: for Research Use Only


Formula: C₆H₁₁NO₃
Boiling Pt: 151 °C (760 mm Hg)
Flash Pt: 72.4 °C
Storage Temperature: Freezer
MDL Number: MFCD06797654
CAS Number: 816-27-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty