Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-ethylhexanoate 95%

Supplier: AMBEED, INC
Warning

A305480-1G A305480-5G A305480-100G A305480-250MG A305480-1G A305480-5G
103814-432EA 31 USD InStock
103814-432 103835-598 76827-934 76827-936 77305-908 77305-910
Ethyl 2-ethylhexanoate 95%
Ethyl 2-ethylhexanoate
Precaución: for Research Use Only


Formula: C₁₀H₂₀O₂
Boiling Pt: 195.7 °C (760 mm Hg)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00462678
CAS Number: 2983-37-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty