Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-benzoylpropionate 95%

Supplier: AMBEED, INC
Warning

A138571-1G A138571-10G A138571-250MG A138571-1G
103802-630EA 41 USD InStock
103802-630 77272-364 77272-366 77315-670
Ethyl-2-benzoylpropionate 95%
Ethyl-2-benzoylpropionate
Precaución: for Research Use Only


Formula: C₁₂H₁₄O₃
Boiling Pt: 279.2 °C (760 mm Hg)
Flash Pt: 117.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00026882
CAS Number: 10488-87-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty