Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3,5-diamino-1H-pyrazole-4-carboxylate 95%

Supplier: AMBEED, INC
Warning

A203663-1G A203663-5G A203663-100MG A203663-10G A203663-250MG A203663-1G A203663-25G A203663-5G
103817-476EA 45 USD InStock
103817-476 103848-136 76986-528 76986-530 76986-532 77086-266 77086-268 77086-270
Ethyl-3,5-diamino-1H-pyrazole-4-carboxylate 95%
Ethyl-3,5-diamino-1H-pyrazole-4-carboxylate
Precaución: for Research Use Only


Formula: C₆H₁₀N₄O₂
Melting Pt: 189…190 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04070845
CAS Number: 6825-71-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty